4-Bromophenylhydrazinium chloride ≥97%

Supplier: BeanTown Chemical

Synonyms: (4-Bromophenyl)hydrazine hydrochloride, 4-Bromophenylhydrazine hydrochloride, 1-Bromo-4-hydrazinobenzene hydrochloride, 1-(4-Bromophenyl)hydrazine hydrochloride

140150-25G 140150-5G
BT140150-25GEA 197 USD
BT140150-25G BT140150-5G
4-Bromophenylhydrazinium chloride ≥97%
4-Bromophenylhydrazinium chloride

CAS: 622-88-8; EC No: 255-590-4; MDL No: MFCD00012941; RTECS: MV0800000 Powder; Molecular Formula: C6H7BrN2·HCl; MW: 223.51 Melting Point: 220-230° (decomposes)


  • RTECS: MV0800000
  • Powder

Formula: C₆H₈BrClN₂
MW: 223.50 g/mol
Melting Pt: 220…230 °C (dec.)
MDL Number: MFCD00012941
CAS Number: 622-88-8
EINECS: 255-590-4

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR