1-Ethyl-1-methylpyrrolidinium bromide ≥99%

Supplier: BeanTown Chemical

Synonyms: 1-Ethyl-1-methylpyrrolidin-1-ium bromide

140135-50G 140135-5G
BT140135-50GEA 138.29 USD
BT140135-50G BT140135-5G
1-Ethyl-1-methylpyrrolidinium bromide ≥99%
1-Ethyl-1-methylpyrrolidinium bromide

CAS: 69227-51-6; EC No: 418-200-5; MDL No: MFCD03095384 Crystalline/Solid; Molecular Formula: C7H16BrN; MW: 194.11


  • Crystalline/Solid

Formula: C₇H₁₆BrN
MW: 194.12 g/mol
MDL Number: MFCD03095384
CAS Number: 69227-51-6
EINECS: 418-200-5

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR