Calcium metasilicate

Supplier: BeanTown Chemical

Synonyms: Calcium silicate

140110-100G 140110-2KG 140110-500G
BT140110-100GEA 45.99 USD
BT140110-100G BT140110-2KG BT140110-500G
Calcium metasilicate
Calcium metasilicate

CAS: 10101-39-0; EC No: 233-250-6; MDL No: MFCD00015979 Powder, typically 20 micron; Linear Formula: CaSiO3; MW: 116.16


  • Powder, typically <20 micron

Formula: CaO₃Si
MW: 116.16 g/mol
MDL Number: MFCD00015979
CAS Number: 10101-39-0
EINECS: 233-250-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR