Antimony ≥99.9999%, polycrystalline lumps 2-8 mm (0.08-0.3 in)
Supplier: BeanTown Chemical
140105-10G
140105-50G
BT140105-10GEA
74
USD
BT140105-10G
BT140105-50G
Antimony ≥99.9999%, polycrystalline lumps 2-8 mm (0.08-0.3 in)
Antimony
CAS: 7440-36-0; EC No: 231-146-5; MDL No: MFCD00134030; RTECS: CC4025000
Solid; Molecular Formula: Sb; MW: 121.76
Melting Point: 630°; Boiling Point: 1635°
Density (g/mL): 6.69
- RTECS: CC4025000
- Solid
Formula:
Sb Boiling Pt: 1635 °C Melting Pt: 630 °C Density: 6.69 g/mL |
MDL Number:
MFCD00134030 CAS Number: 7440-36-0 EINECS: 231-146-5 UN: 2871 ADR: 6.1,III |
Specification Test Results
Assay | 99.9999% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...