Ethyl-2-methylnicotinate ≥97%
Supplier: BeanTown Chemical
Synonyms:
Ethyl 2-methylnicotinate
140090-1G
140090-25G
140090-5G
BT140090-1GEA
25
USD
BT140090-1G
BT140090-25G
BT140090-5G
Ethyl-2-methylnicotinate ≥97%
Ethyl 2-methylnicotinate
CAS: 1721-26-2; EC No: 217-013-4; MDL No: MFCD00006336
Liquid; Molecular Formula: C9H11NO2; MW: 165.19
Boiling Point: 126-127°/24 mmHg; Flash point: 103°C (217°F)
Density (g/mL): 1.072; Refractive Index: 1.505
- Refractive index: 1.505
- Liquid
Formula:
C₉H₁₁NO₂ Boiling Pt: 126…127 °C/24 mmHg Density: 1.072 g/mL Flash Pt: 103 °C (217 °F) |
MDL Number:
MFCD00006336 CAS Number: 1721-26-2 EINECS: 217-013-4 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...