(R)-(-)-2-Amino-3-methyl-1-butanol ≥98%

Supplier: BeanTown Chemical

Synonyms: D(-)-2-Amino-3-methyl-1-butanol, (R)-(-)-2-Amino-3-methylbutanol, D-Valinol, D(-)-Valinol

140045-1G 140045-5G
BT140045-1GEA 61 USD
BT140045-1G BT140045-5G
(R)-(-)-2-Amino-3-methyl-1-butanol ≥98%
(R)-(-)-2-Amino-3-methyl-1-butanol

CAS: 4276-09-9; MDL No: MFCD00064297 Crystalline; Molecular Formula: C5H13NO; MW: 103.16 Melting Point: 35-36°; Flash point: 78°C (172°F) Density (g/mL): 0.931; Refractive Index: 1.455; Optical Rotation: [α]20/D -10±1°, c = 10% in water Air Sensitive


  • Optical rotation: [α]20/D -10±1°, c = 10% in water
  • Refractive index: 1.455
  • Air Sensitive
  • Crystalline

Formula: C₅H₁₃NO
MW: 103.16 g/mol
Melting Pt: 35…36 °C
Density: 0.931 g/mL
Flash Pt: 78 °C (172 °F)
MDL Number: MFCD00064297
CAS Number: 4276-09-9

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR