4-Cyanophenylboronic acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 4-Cyanobenzeneboronic acid, (4-Cyanophenyl)metaboric acid, Dihydroxy(4-cyanophenyl)borane

140040-1G 140040-5G
BT140040-1GEA 66 USD
BT140040-1G BT140040-5G
4-Cyanophenylboronic acid ≥98%
4-Cyanophenylboronic acid

CAS: 126747-14-6; MDL No: MFCD01318968 Powder; Molecular Formula: C7H6BNO2; MW: 146.94 Melting Point: 300°


  • Powder

Formula: C₇H₆BNO₂
MW: 146.94 g/mol
Melting Pt: >300 °C
MDL Number: MFCD01318968
CAS Number: 126747-14-6

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR