Sodium methanesulfonate ≥98%

Supplier: BeanTown Chemical

140030-25G 140030-5G
BT140030-25GEA 468 USD
BT140030-25G BT140030-5G
Sodium methanesulfonate ≥98%
Sodium methanesulphonate

CAS: 2386-57-4; EC No: 219-203-2; MDL No: MFCD00064389 Crystalline; Linear Formula: CH3SO3Na; MW: 118.09 Hygroscopic


  • Hygroscopic
  • Crystalline

Formula: CH₃NaO₃S
MW: 118.09 g/mol
MDL Number: MFCD00064389
CAS Number: 2386-57-4
EINECS: 219-203-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR