Methyl-4-hydroxybenzoate ≥99%
Supplier: BeanTown Chemical
140025-1KG
140025-250G
140025-5KG
BT140025-1KGEA
98.64
USD
BT140025-1KG
BT140025-250G
BT140025-5KG
Methyl-4-hydroxybenzoate ≥99%
Methyl-4-hydroxybenzoate
CAS: 99-76-3; EC No: 202-785-7; MDL No: MFCD00002352; RTECS: DH2450000
Crystalline; Molecular Formula: C8H8O3; MW: 152.15
Melting Point: 125-128°; Boiling Point: 280° (decomposes)
Density (g/mL): 1.88
- RTECS: DH2450000
- Crystalline
Formula:
HOC₆H₄CO₂CH₃ Boiling Pt: 280 °C (dec.) Melting Pt: 125…128 °C Density: 1.88 g/mL |
MDL Number:
MFCD00002352 CAS Number: 99-76-3 EINECS: 202-785-7 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...