Eugenol ≥99%

Supplier: BeanTown Chemical

Synonyms: 4-Allyl-2-methoxyphenol, 5-Allyl-2-hydroxyanisole

140020-100G 140020-500G
BT140020-100GEA 45.88 USD
BT140020-100G BT140020-500G
Eugenol ≥99%
Eugenol

CAS: 97-53-0; EC No: 202-589-1; MDL No: MFCD00008654; RTECS: SJ4375000 Liquid; Molecular Formula: C10H12O2; MW: 164.21 Melting Point: -10°; Boiling Point: 254°; Flash point: 127°C (260°F) Density (g/mL): 1.037; Refractive Index: 1.541


  • RTECS: SJ4375000
  • Refractive index: 1.541
  • Liquid

Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 254 °C
Melting Pt: –10 °C
Density: 1.037 g/mL
Flash Pt: 127 °C (260 °F)
MDL Number: MFCD00008654
CAS Number: 97-53-0
EINECS: 202-589-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR