1,3-Diisopropylbenzene ≥96%

Supplier: BeanTown Chemical

140015-250G 140015-50G
BT140015-250GEA 57 USD
BT140015-250G BT140015-50G
1,3-Diisopropylbenzene ≥96%
1,3-Diisopropylbenzene

CAS: 99-62-7; EC No: 202-773-1; MDL No: MFCD00008889; RTECS: CZ6334000 Liquid; Molecular Formula: C12H18; MW: 162.27 Melting Point: -63°; Boiling Point: 203°; Flash point: 77°C (171°F) Density (g/mL): 0.856; Refractive Index: 1.488


  • RTECS: CZ6334000
  • Refractive index: 1.488
  • Liquid

Formula: C₁₂H₁₈
MW: 162.27 g/mol
Boiling Pt: 203 °C
Melting Pt: –63 °C
Density: 0.856 g/mL
Flash Pt: 77 °C (171 °F)
MDL Number: MFCD00008889
CAS Number: 99-62-7
EINECS: 202-773-1
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR