3,4-Dibromothiophene ≥98%

Supplier: BeanTown Chemical

139935-1G 139935-25G 139935-5G
BT139935-1GEA 28 USD
BT139935-1G BT139935-25G BT139935-5G
3,4-Dibromothiophene ≥98%
3,4-Dibromothiophene

CAS: 3141-26-2; EC No: 221-546-8; MDL No: MFCD00005465 Liquid; Molecular Formula: C4H2Br2S; MW: 241.93 Melting Point: 4-5°; Boiling Point: 221-222°; Flash point: 110°C (230°F) Density (g/mL): 2.188; Refractive Index: 1.640


  • Refractive index: 1.640
  • Liquid

Formula: C₄H₂Br₂S
MW: 241.93 g/mol
Boiling Pt: 221…222 °C
Melting Pt: 4…5 °C
Density: 2.188 g/mL
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00005465
CAS Number: 3141-26-2
EINECS: 221-546-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR