1-Boc-4-hydroxypiperidine ≥98%

Supplier: BeanTown Chemical

Synonyms: N-Boc-4-piperidinol, tert-butyl 4-hydroxypiperidine-1-carboxylate, N-BOC-4-Hydroxypiperidine, tert-Butyl 4-hydroxypiperidine-1-carboxylate

139905-1G 139905-5G
BT139905-1GEA 25 USD
BT139905-1G BT139905-5G
1-Boc-4-hydroxypiperidine ≥98%
1-Boc-4-hydroxypiperidine

CAS: 109384-19-2; MDL No: MFCD01075174 Solid; Molecular Formula: C10H19NO3; MW: 201.27 Melting Point: 61-65°


  • Solid

Formula: C₁₀H₁₉NO₃
MW: 201.27 g/mol
Melting Pt: 61…65 °C
MDL Number: MFCD01075174
CAS Number: 109384-19-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR