Sodium dihydrogen phosphate ≥96%
Supplier: BeanTown Chemical
Synonyms:
Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic
139880-1KG
139880-250G
BT139880-1KGEA
59.81
USD
BT139880-1KG
BT139880-250G
Sodium dihydrogen phosphate ≥96%
Sodium dihydrogen phosphate
CAS: 7558-80-7; EC No: 231-449-2; MDL No: MFCD00003527; RTECS: WA1900000
Powder; Linear Formula: NaH2PO4; MW: 119.98
Hygroscopic
- RTECS: WA1900000
- Hygroscopic
- Powder
Formula:
NaH₂PO₄ MW: 119.98 g/mol |
MDL Number:
MFCD00003527 CAS Number: 7558-80-7 EINECS: 231-449-2 |
Specification Test Results
Assay | 96% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...