Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate ≥85%, Technical Grade
Supplier: BeanTown Chemical
139875-1G
139875-250MG
139875-5G
BT139875-1GEA
288
USD
BT139875-1G
BT139875-250MG
BT139875-5G
Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate ≥85%, Technical Grade
Triphenylphosphine-3,3',3''-trisulfonic acid trisodium salt hydrate
CAS: 63995-70-0; EC No: 264-596-6; MDL No: MFCD00145472
Crystalline; Linear Formula: P(C6H4SO3Na)3·xH2O ; MW: 568.41 (anhydrous)
- Crystalline
Formula:
C₁₈H₁₂Na₃O₉PS₃.xH₂O MW: 568.408 g/mol (anhy) |
MDL Number:
MFCD11046357 CAS Number: 63995-70-0 EINECS: 264-596-6 |
Specification Test Results
Assay | 85% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...