4-Fluoro-2-iodotoluene ≥98%
Supplier: BeanTown Chemical
139830-25G
139830-5G
BT139830-25GEA
294
USD
BT139830-25G
BT139830-5G
4-Fluoro-2-iodotoluene ≥98%
4-Fluoro-2-iodotoluene
CAS: 13194-67-7; EC No: 236-153-7; MDL No: MFCD00013709
Liquid; Molecular Formula: C7H6FI; MW: 236.03
Boiling Point: 92-94°/15 mmHg; Flash point: 86°C (187°F)
Density (g/mL): 1.752; Refractive Index: 1.580
Light Sensitive
- Refractive index: 1.580
- Light Sensitive
- Liquid
Formula:
C₇H₆FI Boiling Pt: 92…94 °C/15 mmHg Density: 1.752 g/mL Flash Pt: 86 °C (187 °F) |
MDL Number:
MFCD00013709 CAS Number: 13194-67-7 EINECS: 236-153-7 UN: 1993 ADR: 3,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...