Iron disilicide ≥99.9% (trace metals basis)

Supplier: BeanTown Chemical

139765-25G 139765-5G
BT139765-25GEA 68 USD
BT139765-25G BT139765-5G
Iron disilicide ≥99.9% (trace metals basis)
Iron disilicide

CAS: 12022-99-0; EC No: 234-671-8; MDL No: MFCD00016087 -20 Mesh Powder; Molecular Formula: FeSi2; MW: 112.02 Melting Point: 1220°


  • -20 Mesh Powder

Formula: FeSi₂
Melting Pt: 1220 °C
MDL Number: MFCD00016087
CAS Number: 12022-99-0
EINECS: 234-671-8

Order Now

Specification Test Results

Assay 99.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR