Calcium sulfate dihydrate 98.0-102.0% ACS

Supplier: BeanTown Chemical

Synonyms: Plaster of Paris

139735-1KG 139735-250G
BT139735-1KGEA 147.08 USD
BT139735-1KG BT139735-250G
Calcium sulfate dihydrate 98.0-102.0% ACS
Calcium sulphate dihydrate

CAS: 10101-41-4; EC No: 231-900-3; MDL No: MFCD00149625; RTECS: EW4150000 Powder; Linear Formula: CaSO4·2H2O; MW: 172.17 Hygroscopic


  • RTECS: EW4150000
  • Hygroscopic
  • Powder

Formula: CaSO₄·2H₂O
MW: 172.17 g/mol
MDL Number: MFCD00149625
CAS Number: 10101-41-4
EINECS: 231-900-3

Order Now

Specification Test Results

Assay 98.0-102.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR