Ethyl-2-pentynoate ≥98%

Supplier: BeanTown Chemical

Synonyms: Ethyl pent-2-ynoate

139725-25G 139725-5G
BT139725-25GEA 311 USD
BT139725-25G BT139725-5G
Ethyl-2-pentynoate ≥98%
Ethyl-2-pentynoate

CAS: 55314-57-3; EC No: 259-588-4; MDL No: MFCD00015221 Liquid; Molecular Formula: C7H10O2; MW: 126.16 Boiling Point: 178-179°; Flash point: 38°C (100°F) Density (g/mL): 0.957; Refractive Index: 1.439


  • Refractive index: 1.439
  • Liquid

Formula: C₇H₁₀O₂
MW: 126.155 g/mol
Boiling Pt: 178…179 °C
Density: 0.957 g/mL
Flash Pt: 38 °C (100 °F)
MDL Number: MFCD00015221
CAS Number: 55314-57-3
EINECS: 259-588-4

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR