Vitamin K1 (phytomenadione, phylloquinone)
Supplier: BeanTown Chemical
Synonyms:
2-Methyl-3-((7R,11R,E)-3,7,11,15-tetramethylhexadec-2-en-1-yl)naphthalene-1,4-dione
139705-1G
139705-5G
BT139705-1GEA
54.83
USD
BT139705-1G
BT139705-5G
Vitamin K1 (phytomenadione, phylloquinone)
Vitamin K1 (phytomenadione, phylloquinone)
CAS: 84-80-0; EC No: 201-564-2; MDL No: MFCD00214063; RTECS: QJ5800000
Liquid; Molecular Formula: C31H46O2; MW: 450.70
Melting Point: -20°; Boiling Point: 140°/0.001 mmHg; Flash point:
- RTECS: QJ5800000
- Refractive index: 1.527
- Air Sensitive, Light Sensitive, Moisture Sensitive
- Liquid
Formula:
C₃₁H₄₆O₂ Boiling Pt: 140 °C/0.001 mmHg Melting Pt: –20 °C Density: 0.984 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00214063 CAS Number: 84-80-0 EINECS: 201-564-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...