trans-4-(4-n-Pentylcyclohexyl)phenyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate ≥97%
Supplier: BeanTown Chemical
Synonyms:
4-(trans-4-Pentylcyclohexyl)phenyl (trans,trans)-4'-Propyl-[1,1'-bi(cyclohexane)]-4-carboxylate, (trans,trans)-4'-Propyl-[1,1'-bi(cyclohexane)]-4-carboxylic acid 4-(trans-4-Pentylcyclohexyl)phenyl ester
139700-1G
139700-5G
BT139700-1GEA
77
USD
BT139700-1G
BT139700-5G
trans-4-(4-n-Pentylcyclohexyl)phenyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate ≥97%
trans-4-(4-n-Pentylcyclohexyl)phenyl-trans,trans-4'-n-propylbicyclohexyl-4-carboxylate
CAS: 131790-57-3; MDL No: MFCD11053459
Solid; Molecular Formula: C31H48O2; MW: 452.72
Melting Point: 213-223°
- Solid
Formula:
C₃₃H₅₂O₂ MW: 480.777 g/mol Melting Pt: 213…223 °C |
MDL Number:
MFCD11053459 CAS Number: 131790-57-3 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...