Sodium metatungstate monohydrate

Supplier: BeanTown Chemical

139685-250G 139685-50G
BT139685-250GEA 373.84 USD
BT139685-250G BT139685-50G
Sodium metatungstate monohydrate
Sodium metatungstate monohydrate

CAS: 12141-67-2; EC No: 412-770-9; MDL No: MFCD00061466 Powder; Linear Formula: Na6W12O39·H2O; MW: 2986.12


  • Powder

Formula: Na₆O₃₉W₁₂·H₂O
MDL Number: MFCD00061466
CAS Number: 12141-67-2
EINECS: 412-770-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR