(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one ≥98%

Supplier: BeanTown Chemical

Synonyms: 2-azabicyclo[2.2.1]hept-5-en-3-one

139655-1G 139655-250MG 139655-5G
BT139655-1GEA 74 USD
BT139655-1G BT139655-250MG BT139655-5G
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one ≥98%
(±)-2-Azabicyclo[2.2.1]hept-5-en-3-one

CAS: 49805-30-3; EC No: 421-830-3; MDL No: MFCD00213364 Powder; Molecular Formula: C6H7NO; MW: 109.13 Melting Point: 54-58°; Boiling Point: 102-106°/0.25 mmHg; Flash point: 110°C (230°F)


  • Powder

Formula: C₆H₇NO
Boiling Pt: 102…106 °C/0.25 mmHg
Melting Pt: 54…58 °C
Flash Pt: 110 °C (230 °F)
MDL Number: MFCD00213364
CAS Number: 49805-30-3
EINECS: 421-830-3

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR