1-Dodecene ≥96%

Supplier: BeanTown Chemical

Synonyms: Dodec-1-ene

139635-100G 139635-500G
BT139635-100GEA 64.94 USD
BT139635-100G BT139635-500G
1-Dodecene ≥96%
1-Dodecene

CAS: 112-41-4; EC No: 203-968-4; MDL No: MFCD00008961 Liquid; Linear Formula: CH3(CH2)9CH=CH2; Molecular Formula: C12H24; MW: 168.32 Melting Point: -35°; Boiling Point: 214-216°; Flash point: 73°C (163°F) Density (g/mL): 0.758; Refractive Index: 1.429


  • Refractive index: 1.429
  • Liquid

Formula: C₁₂H₂₄
MW: 168.32 g/mol
Boiling Pt: 214…216 °C
Melting Pt: –35 °C
Density: 0.758 g/mL
Flash Pt: 73 °C (163 °F)
MDL Number: MFCD00008961
CAS Number: 112-41-4
EINECS: 203-968-4

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR