Titanium nitride ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

139620-1KG 139620-250G 139620-50G
BT139620-1KGEA 638 USD
BT139620-1KG BT139620-250G BT139620-50G
Titanium nitride ≥99.5% (trace metals basis)
Titanium nitride

CAS: 25583-20-4; EC No: 247-117-5; MDL No: MFCD00049596; RTECS: XR2230000 10 Micron Powder; Linear Formula: TiN; MW: 61.89 Melting Point: 2930° Density (g/mL): 5.24


  • RTECS: XR2230000
  • <10 Micron Powder

Formula: NTi
Melting Pt: 2930 °C
Density: 5.24 g/mL
MDL Number: MFCD00049596
CAS Number: 25583-20-4
EINECS: 247-117-5

Order Now

Specification Test Results

Assay 99.5%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR