Tin ≥99.999% (trace metals basis), shot 3 mm
Supplier: BeanTown Chemical
139600-100G
139600-25G
139600-500G
BT139600-100GEA
357.02
USD
BT139600-100G
BT139600-25G
BT139600-500G
Tin ≥99.999% (trace metals basis), shot 3 mm
Tin
CAS: 7440-31-5; EC No: 231-141-8; MDL No: MFCD00133862; RTECS: XP7320000
Solid; Molecular Formula: Sn; MW: 118.71
Melting Point: 231.9°; Boiling Point: 2270°
Density (g/mL): 7.31
- RTECS: XP7320000
- Solid
Formula:
Sn MW: 118.71 g/mol Boiling Pt: 2270 °C Melting Pt: 231.9 °C Density: 7.31 g/mL |
MDL Number:
MFCD00133862 CAS Number: 7440-31-5 EINECS: 231-141-8 |
Specification Test Results
Assay | 99.999% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...