Potassium oxalate hydrate 99%

Supplier: BeanTown Chemical
Warning

Synonyms: Oxalic acid potassium salt hydrate, Ethanedioic acid potassium salt

139590-1KG 139590-250G 139590-5KG
BT139590-1KGEA 97.27 USD
BT139590-1KG BT139590-250G BT139590-5KG
Potassium oxalate hydrate 99%
Potassium oxalate hydrate

CAS: 6487-48-5; EC No: 209-506-8; MDL No: MFCD00150033; RTECS: RO2885000 Powder; Linear Formula: K2C2O4·xH2O; MW: 166.22 (anhydrous) Melting Point: 300° Density (g/mL): 2.127 Hygroscopic


  • RTECS: RO2885000
  • Hygroscopic
  • Powder

Formula: (COOK)₂·xH₂O
MW: 166.22 g/mol (anhy)
Melting Pt: >300 °C
Density: 2.127 g/mL
MDL Number: MFCD00150033
CAS Number: 6487-48-5
EINECS: 209-506-8
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR