3,4-Difluorobenzaldehyde ≥98%
Supplier: BeanTown Chemical
139555-25G
139555-5G
BT139555-25GEA
181
USD
BT139555-25G
BT139555-5G
3,4-Difluorobenzaldehyde ≥98%
3,4-Difluorobenzaldehyde
CAS: 34036-07-2; EC No: 422-180-3; MDL No: MFCD00010328
Liquid; Molecular Formula: C7H4F2O; MW: 142.10
Flash point: 66°C (151°F)
Density (g/mL): 1.288; Refractive Index: 1.500
Air Sensitive
- Refractive index: 1.500
- Air Sensitive
- Liquid
Formula:
C₇H₄F₂O MW: 142.1 g/mol Density: 1.288 g/mL Flash Pt: 66 °C (151 °F) |
MDL Number:
MFCD00010328 CAS Number: 34036-07-2 EINECS: 422-180-3 |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...