1,2,4-Triaminobenzene dihydrochloride ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,2,4-Benzenetriamine Dihydrochloride
139545-25G
139545-5G
BT139545-25GEA
347
USD
BT139545-25G
BT139545-5G
1,2,4-Triaminobenzene dihydrochloride ≥98%
1,2,4-Triaminobenzene dihydrochloride
CAS: 615-47-4; EC No: 210-428-1; MDL No: MFCD00016619; RTECS: DC1953000
Solid; Molecular Formula: C6H9N3·2HCl ; MW: 196.08
Hygroscopic
- RTECS: DC1953000
- Hygroscopic
- Solid
Formula:
C₆H₉N₃ MW: 196.08 g/mol |
MDL Number:
MFCD00016619 CAS Number: 615-47-4 EINECS: 210-428-1 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...