Cobalt (III) acetylacetonate

Supplier: BeanTown Chemical

Synonyms: Cobalt (III) tris(2,4-pentanedionato-O,O'), Cobalt (III) 2,4-pentanedionate, Cobalt tris(acetylacetonate), Tris(2,4-pentanedionato-O,O')cobalt (III)

139540-100G 139540-25G 139540-5G
BT139540-100GEA 134.63 USD
BT139540-100G BT139540-25G BT139540-5G
Cobalt (III) acetylacetonate
Cobalt (III) acetylacetonate

CAS: 21679-46-9; EC No: 244-527-6; MDL No: MFCD00013488 Powder; Molecular Formula: C15H21CoO6 ; MW: 356.26 Melting Point: 210-213° Hygroscopic, Moisture Sensitive


  • Hygroscopic, Moisture Sensitive
  • Powder

Formula: C₁₅H₂₁CoO₆
MW: 356.26 g/mol
Melting Pt: 210…213 °C
MDL Number: MFCD00013488
CAS Number: 21679-46-9
EINECS: 244-527-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR