Benzo[b]thiophene-3-carboxylic acid ≥96%

Supplier: BeanTown Chemical

Synonyms: 1-Benzothiophene-3-carboxylic acid

139520-1G 139520-25G 139520-5G
BT139520-1GEA 97 USD
BT139520-1G BT139520-25G BT139520-5G
Benzo[b]thiophene-3-carboxylic acid ≥96%
Benzo[b]thiophene-3-carboxylic acid

CAS: 5381-25-9; MDL No: MFCD01846406 Solid; Molecular Formula: C9H6O2S; MW: 178.22 Melting Point: 174-177°


  • Solid

Formula: C₉H₆O₂S
MW: 178.22 g/mol
Melting Pt: 174…177 °C
MDL Number: MFCD01846406
CAS Number: 5381-25-9

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR