Benzyltrimethylammonium dichloroiodate ≥95%
Supplier: BeanTown Chemical
139490-25G
139490-5G
BT139490-25GEA
182
USD
BT139490-25G
BT139490-5G
Benzyltrimethylammonium dichloroiodate ≥95%
Benzyltrimethylammonium dichloroiodate
CAS: 114971-52-7; MDL No: MFCD00075259
Crystalline; Molecular Formula: C10H16NICl2 ; MW: 348.05
Melting Point: 124-126°
Hygroscopic , Light Sensitive
- Hygroscopic, Light Sensitive
- Crystalline
Formula:
C₁₀H₁₆Cl₂IN MW: 348.06 g/mol Melting Pt: 124…126 °C |
MDL Number:
MFCD00075259 CAS Number: 114971-52-7 |
Specification Test Results
Assay | 95% |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...