Dirhodium(II) tetrakis (caprolactam)
Supplier: BeanTown Chemical
139475-0.1G
139475-25MG
139475-500MG
BT139475-0.1GEA
300.34
USD
BT139475-0.1G
BT139475-25MG
BT139475-500MG
Dirhodium(II) tetrakis (caprolactam)
Dirhodium(II) tetrakis (caprolactam)
CAS: 138984-26-6; MDL No: MFCD00467690
Powder; Molecular Formula: C24H40N4O4Rh2; MW: 736.52
Melting Point: 269-274°
- Powder
Formula:
C₂₄H₄₀N₄O₄Rh₂ Melting Pt: 269…274 °C |
MDL Number:
MFCD00467690 CAS Number: 138984-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...