(R)-tert-Butyl(2-oxooxetan-3-yl)carbamate ≥95%
Supplier: BeanTown Chemical
Synonyms:
N-Boc-D-serine-β-lactone, (R)-tert-Butyl (2-oxooxetan-3-yl)carbamate
139465-1G
139465-250MG
BT139465-1GEA
479
USD
BT139465-1G
BT139465-250MG
(R)-tert-Butyl(2-oxooxetan-3-yl)carbamate ≥95%
(R)-tert-Butyl(2-oxooxetan-3-yl)carbamate
CAS: 126330-77-6
Solid; Molecular Formula: C8H13NO4; MW: 187.19
Melting Point: 121-122°; Boiling Point: 319°; Flash point: 147°C (297°F)
Density (g/mL): 1.185
- Solid
Formula:
C₈H₁₃NO₄ Boiling Pt: 319 °C Melting Pt: 121…122 °C Density: 1.185 g/mL Flash Pt: 147 °C (297 °F) |
MDL Number:
MFCD11519129 CAS Number: 126330-77-6 |
Specification Test Results
Assay | 95% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...