Lanthanum(III) chloride heptahydrate ACS

Supplier: BeanTown Chemical

139455-250G 139455-50G
BT139455-250GEA 301.29 USD
BT139455-250G BT139455-50G
Lanthanum(III) chloride heptahydrate ACS
Lanthanum(III) chloride heptahydrate

CAS: 10025-84-0; EC No: 233-237-5; MDL No: MFCD00149756; RTECS: OE4470000 Crystalline; Linear Formula: LaCl3·7H2O; MW: 371.37 Melting Point: 91° (decomposes)


  • RTECS: OE4470000
  • Crystalline

Formula: LaCl₃·7H₂O
MW: 371.37 g/mol
Melting Pt: 91 °C (dec.)
MDL Number: MFCD00149756
CAS Number: 10025-84-0
EINECS: 233-237-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR