D-Glucoronic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
D-(+)Glucuronic acid, D-Glucuronic acid, D-(+)-glucuronic acid, (2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid
139445-25G
139445-5G
BT139445-25GEA
112
USD
BT139445-25G
BT139445-5G
D-Glucoronic acid ≥98%
D-Glucoronic acid
CAS: 6556-12-3; EC No: 229-486-4; MDL No: MFCD00077778; RTECS: LZ8836600
Powder; Molecular Formula: C6H10O7; MW: 194.14
Melting Point: 159-161°
- RTECS: LZ8836600
- Powder
Formula:
C₆H₁₀O₇ Melting Pt: 159…161 °C |
MDL Number:
MFCD00166981 CAS Number: 6556-12-3 EINECS: 229-486-4 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...