9,10-Diphenylanthracene ≥97%

Supplier: BeanTown Chemical

139440-1G 139440-25G 139440-5G
BT139440-1GEA 40 USD
BT139440-1G BT139440-25G BT139440-5G
9,10-Diphenylanthracene ≥97%
9,10-Diphenylanthracene

CAS: 1499-10-1; EC No: 216-105-1; MDL No: MFCD00001253 Powder; Molecular Formula: C26H18; MW: 330.42 Melting Point: 245-248°


  • Powder

Formula: C₂₆H₁₈
MW: 330.43 g/mol
Melting Pt: 245…248 °C
MDL Number: MFCD00001253
CAS Number: 1499-10-1
EINECS: 216-105-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR