Iron nickel oxide, Technical Grade

Supplier: BeanTown Chemical

Synonyms: Nickel ferrite

139340-10G 139340-50G
BT139340-10GEA 49 USD
BT139340-10G BT139340-50G
Iron nickel oxide, Technical Grade
Iron nickel oxide

CAS: 12168-54-6; EC No: 235-335-3; MDL No: MFCD00016254 Powder; Molecular Formula: NiFe2O4; MW: 234.39 Density (g/mL): 5.368


  • Powder

Formula: NiFe₂O₄
MW: 234.39 g/mol
Density: 5.368 g/mL
MDL Number: MFCD00016254
CAS Number: 12168-54-6
EINECS: 235-335-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR