Indium(III) hydroxide ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical

139320-10G 139320-50G
BT139320-10GEA 88 USD
BT139320-10G BT139320-50G
Indium(III) hydroxide ≥99.995% (trace metals basis)
Indium(III) hydroxide

CAS: 20661-21-6; EC No: 243-947-7; MDL No: MFCD00011056 -100 Mesh Powder; Linear Formula: In(OH)3; MW: 165.84 Melting Point: 150° Density (g/mL): 4.45


  • -100 Mesh Powder

Formula: H₃InO₃
MW: 165.84 g/mol
Melting Pt: 150 °C
Density: 4.45 g/mL
MDL Number: MFCD00011056
CAS Number: 20661-21-6
EINECS: 243-947-7

Order Now

Specification Test Results

Assay 99.995%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR