3-Methyl-4-cyclohexene-1,2-dicarboxylic anhydride ≥95%

Supplier: BeanTown Chemical

Synonyms: 3-Methyl-DELTA4-tetrahydrophthalic Anhydride, 3-Methyl-1,2,3,6-tetrahydrophthalic anhydride, 3-Methyl-4-cyclohexene-1,2-dicarboxylic Anhydride

139310-1G 139310-5G
BT139310-1GEA 162 USD
BT139310-1G BT139310-5G
3-Methyl-4-cyclohexene-1,2-dicarboxylic anhydride ≥95%
3-Methyl-4-cyclohexene-1,2-dicarboxylic anhydride

CAS: 5333-84-6; EC No: 226-247-6; MDL No: MFCD00014585 Solid; Molecular Formula: C9H10O3; MW: 166.17


  • Solid

Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD00014585
CAS Number: 5333-84-6
EINECS: 226-247-6

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR