CAPS (3-(cyclohexylamino)propanesulfonic acid) ≥99%

Supplier: BeanTown Chemical

Synonyms: 3-Cyclohexylaminopropanesulphonic acid

139215-100G 139215-25G 139215-500G
BT139215-100GEA 62.97 USD
BT139215-100G BT139215-25G BT139215-500G
CAPS (3-(cyclohexylamino)propanesulfonic acid) ≥99%
CAPS (3-(cyclohexylamino)propanesulphonic acid)

CAS: 1135-40-6; EC No: 214-492-1; MDL No: MFCD00003837; RTECS: TZ6395000 Powder; Molecular Formula: C9H19NO3S; MW: 221.32


  • RTECS: TZ6395000
  • Powder

Formula: C₆H₁₁NH(CH₂)₃SO₃H
MW: 221.32 g/mol
MDL Number: MFCD00003837
CAS Number: 1135-40-6
EINECS: 214-492-1

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR