3,6-Dimethyl-1,4-dioxane-2,5-dione ≥99%

Supplier: BeanTown Chemical

Synonyms: DL-Lactide

139160-10G 139160-50G
BT139160-10GEA 41.51 USD
BT139160-10G BT139160-50G
3,6-Dimethyl-1,4-dioxane-2,5-dione ≥99%
3,6-Dimethyl-1,4-dioxane-2,5-dione

CAS: 95-96-5; EC No: 202-468-3; MDL No: MFCD00011685; RTECS: JH1100000 Powder; Molecular Formula: C6H8O4; MW: 144.13 Melting Point: 116-119°; Boiling Point: 142°/8 mmHg Moisture Sensitive


  • RTECS: JH1100000
  • Moisture Sensitive
  • Powder

Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 142 °C/8 mmHg
Melting Pt: 116…119 °C
MDL Number: MFCD00011685
CAS Number: 95-96-5
EINECS: 202-468-3

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR