trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl ≥97%
Supplier: BeanTown Chemical
Synonyms:
(trans,trans)-4-(3,4-Difluorophenyl)-4'-vinyl-1,1'-bi(cyclohexane)
139155-1G
139155-5G
BT139155-1GEA
103
USD
BT139155-1G
BT139155-5G
trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl ≥97%
trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl
CAS: 142400-92-8; MDL No: MFCD09842838
Solid; Molecular Formula: C20H26F2; MW: 304.42
- Solid
Formula:
C₂₀H₂₆F₂ MW: 304.425 g/mol |
MDL Number:
MFCD09842838 CAS Number: 142400-92-8 |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...