1,2,3-Triazole ≥98%
Supplier: BeanTown Chemical
Synonyms:
1,2,3-1H-Triazole, 1H-1,2,3-Triazole
139145-1G
139145-25G
139145-5G
BT139145-1GEA
61.3
USD
BT139145-1G
BT139145-25G
BT139145-5G
1,2,3-Triazole ≥98%
1,2,3-Triazole
CAS: 288-36-8; MDL No: MFCD00014490
Liquid; Molecular Formula: C2H3N3; MW: 69.07
Melting Point: 23-25°; Boiling Point: 203°/752 mmHg; Flash point: 107°C (225°F)
Density (g/mL): 1.192; Refractive Index: 1.498
- Refractive index: 1.498
- Liquid
Formula:
C₂H₃N₃ Boiling Pt: 203 °C/752 mmHg Melting Pt: 23…25 °C Density: 1.192 g/mL Flash Pt: 107 °C (225 °F) |
MDL Number:
MFCD00014490 CAS Number: 288-36-8 |
Specification Test Results
Assay | 98% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...