Boc-Dab(Fmoc)-OH ≥95%

Supplier: BeanTown Chemical

139140-1G 139140-250MG 139140-5G
BT139140-1GEA 121 USD
BT139140-1G BT139140-250MG BT139140-5G
Boc-Dab(Fmoc)-OH ≥95%
Boc-Dab(Fmoc)-OH

CAS: 117106-21-5; MDL No: MFCD00236845 Powder; Molecular Formula: C24H28N2O6; MW: 440.49


  • Powder

Formula: C₂₄H₂₈N₂O₆
MW: 440.496 g/mol
MDL Number: MFCD00236845
CAS Number: 117106-21-5

Order Now

Specification Test Results

Assay 95%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR