trans-1-[(4-Ethylphenyl)ethylnyl]-4-(4-propylcyclohexyl)benzene ≥97%

Supplier: BeanTown Chemical

Synonyms: 1-Ethyl-4-((4-(trans-4-propylcyclohexyl)phenyl)ethynyl)benzene

139130-1G 139130-250MG
BT139130-1GEA 155 USD
BT139130-1G BT139130-250MG
trans-1-[(4-Ethylphenyl)ethylnyl]-4-(4-propylcyclohexyl)benzene ≥97%
trans-1-[(4-Ethylphenyl)ethylnyl]-4-(4-propylcyclohexyl)benzene

CAS: 107949-21-3; MDL No: MFCD11053405 Powder; Molecular Formula: C25H30; MW: 330.51 Melting Point: 107-109°


  • Powder

Formula: C₂₅H₃₀
MW: 330.515 g/mol
Melting Pt: 107…109 °C
MDL Number: MFCD11053405
CAS Number: 107949-21-3

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR