Potassium acetate ≥99%
Supplier: BeanTown Chemical
139055-10KG
139055-2.5KG
139055-500G
BT139055-10KGEA
456
USD
BT139055-10KG
BT139055-2.5KG
BT139055-500G
Potassium acetate ≥99%
Potassium acetate
CAS: 127-08-2; EC No: 204-822-2; MDL No: MFCD00012458; RTECS: AJ3325000
Crystalline; Linear Formula: CH3COOK; MW: 98.15
Melting Point: 292°
Density (g/mL): 1.57
Hygroscopic
- RTECS: AJ3325000
- Hygroscopic
- Crystalline
Formula:
CH₃COOK Melting Pt: 292 °C Density: 1.57 g/mL |
MDL Number:
MFCD00012458 CAS Number: 127-08-2 EINECS: 204-822-2 |
|
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...