Methanesulfonamide ≥98%

Supplier: BeanTown Chemical

139025-10G 139025-250G 139025-50G
BT139025-10GEA 30.83 USD
BT139025-10G BT139025-250G BT139025-50G
Methanesulfonamide ≥98%
Methanesulphonamide

CAS: 3144-09-0; EC No: 221-553-6; MDL No: MFCD00007940 Crystalline; Linear Formula: CH3SO2NH2; MW: 95.12 Melting Point: 87-91°


  • Crystalline

Formula: CH₅NO₂S
Melting Pt: 87…91 °C
MDL Number: MFCD00007940
CAS Number: 3144-09-0
EINECS: 221-553-6

Order Now

Specification Test Results

Assay 98%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR