5-Bromo-2-furoic acid ≥99%

Supplier: BeanTown Chemical

139020-250G 139020-50G
BT139020-250GEA 296 USD
BT139020-250G BT139020-50G
5-Bromo-2-furoic acid ≥99%
5-Bromo-2-furoic acid

CAS: 585-70-6; EC No: 209-559-7; MDL No: MFCD00003239 Crystalline/Powder; Molecular Formula: C5H3BrO3; MW: 190.98 Melting Point: 188-190°


  • Crystalline/Powder

Formula: C₅H₃BrO₃
Melting Pt: 188…190 °C
MDL Number: MFCD00003239
CAS Number: 585-70-6
EINECS: 209-559-7

Order Now

Specification Test Results

Assay 99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR