2-Thiobarbituric acid ≥98%

Supplier: BeanTown Chemical

Synonyms: 4,6-Dihydroxy-2-mercaptopyrimidine

139015-100G 139015-25G 139015-500G
BT139015-100GEA 113.66 USD
BT139015-100G BT139015-25G BT139015-500G
2-Thiobarbituric acid ≥98%
2-Thiobarbituric acid

CAS: 504-17-6; EC No: 207-985-8; MDL No: MFCD00006674; RTECS: CQ7700000 Powder; Molecular Formula: C4H4N2O2S; MW: 144.15 Melting Point: 245° (decomposes)


  • RTECS: CQ7700000
  • Powder

Formula: C₄H₄N₂O₂S
MW: 144.15 g/mol
Melting Pt: 245 °C (dec.)
MDL Number: MFCD00006674
CAS Number: 504-17-6
EINECS: 207-985-8

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR